SY3H3D680C7A Synthonix Corporation

7-Methoxy-4H-chromen-4-one

  • Cas Number : 5751-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53807.html

Safety Information

MSDS https://synthonix.com/53807.html
Availability details for
Quantity
  • Availability
  • Add to Cart