SY3H3D680C80 Synthonix Corporation

Methyl 6-chloro-4-methoxy-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 871583-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53864.html

Safety Information

MSDS https://synthonix.com/53864.html
Availability details for
Quantity
  • Availability
  • Add to Cart