SY3H3D680C84 Synthonix Corporation

4-(5-Methyl-1H-1,2,4-triazol-1-yl)benzoic acid

  • MDL number No MDL found
  • Cas Number : 1368819-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53880.html

Safety Information

MSDS https://synthonix.com/53880.html
Availability details for
Quantity
  • Availability
  • Add to Cart