SY3H3D680C85 Synthonix Corporation

Methyl 3-(pyridin-2-yl)-1H-pyrazole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 1340553-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53882.html

Safety Information

MSDS https://synthonix.com/53882.html
Availability details for
Quantity
  • Availability
  • Add to Cart