SY3H3D680C88 Synthonix Corporation

5-Methoxyquinoline

  • MDL number No MDL found
  • Cas Number : 6931-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/53891.html

Safety Information

MSDS https://synthonix.com/53891.html
Availability details for
Quantity
  • Availability
  • Add to Cart