SY3H3D680C8B Synthonix Corporation

4-Methylquinolin-7-ol

  • MDL number No MDL found
  • Cas Number : 15463-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53895.html

Safety Information

MSDS https://synthonix.com/53895.html
Availability details for
Quantity
  • Availability
  • Add to Cart