SY3H3D680C8F Synthonix Corporation

6-Methylquinolin-5-ol

  • MDL number No MDL found
  • Cas Number : 116529-84-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/53899.html

Safety Information

MSDS https://synthonix.com/53899.html
Availability details for
Quantity
  • Availability
  • Add to Cart