SY3H3D680C90 Synthonix Corporation

6-Methylquinolin-8-ol

  • MDL number No MDL found
  • Cas Number : 20984-33-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53900.html

Safety Information

MSDS https://synthonix.com/53900.html
Availability details for
Quantity
  • Availability
  • Add to Cart