SY3H3D680C98 Synthonix Corporation

1-Methyl-1H-indole-7-carbaldehyde

  • Cas Number : 69047-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53911.html

Safety Information

MSDS https://synthonix.com/53911.html
Availability details for
Quantity
  • Availability
  • Add to Cart