SY3H3D680C9E Synthonix Corporation

6-Methoxyisoquinolin-3(2H)-one

  • Cas Number : 51463-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53928.html

Safety Information

MSDS https://synthonix.com/53928.html
Availability details for
Quantity
  • Availability
  • Add to Cart