SY3H3D680C9F Synthonix Corporation

Methyl 3-(cyanomethyl)benzoate

  • MDL number MFCD07783720
  • Cas Number : 68432-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown to Red Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/53929.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/53929.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart