SY3H3D680CA5 Synthonix Corporation

Methyl indolizine-7-carboxylate

  • MDL number No MDL found
  • Cas Number : 887602-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53937.html

Safety Information

MSDS https://synthonix.com/53937.html
Availability details for
Quantity
  • Availability
  • Add to Cart