SY3H3D680CA7 Synthonix Corporation

1-Methyl-1H-indole-6-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 202745-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53940.html

Safety Information

MSDS https://synthonix.com/53940.html
Availability details for
Quantity
  • Availability
  • Add to Cart