SY3H3D680CB2 Synthonix Corporation

Methyl (4-Cyanophenoxy)acetate

  • MDL number No MDL found
  • Cas Number : 272792-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53955.html

Safety Information

MSDS https://synthonix.com/53955.html
Availability details for
Quantity
  • Availability
  • Add to Cart