SY3H3D680CB8 Synthonix Corporation

Methyl 5-methylbenzo[d]oxazole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 27383-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53967.html

Safety Information

MSDS https://synthonix.com/53967.html
Availability details for
Quantity
  • Availability
  • Add to Cart