SY3H3D680D09 Synthonix Corporation

2-Methyl-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-6-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1092352-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/53980.html

Safety Information

MSDS https://synthonix.com/53980.html
Availability details for
Quantity
  • Availability
  • Add to Cart