SY3H3D680D0B Synthonix Corporation

Methyl 3-(4-bromophenyl)acrylate

  • MDL number No MDL found
  • Cas Number : 3650-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/53987.html

Safety Information

MSDS https://synthonix.com/53987.html
Availability details for
Quantity
  • Availability
  • Add to Cart