SY3H3D680D0D Synthonix Corporation

Methyl 2-methyl-3-(trifluoromethyl)benzoate

  • MDL number MFCD14698066
  • Cas Number : 346603-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54014.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54014.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart