SY3H3D680D0F Synthonix Corporation

1-(4-Methoxy-2-(trifluoromethyl)phenyl)ethanone

  • MDL number No MDL found
  • Cas Number : 220141-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54016.html

Safety Information

MSDS https://synthonix.com/54016.html
Availability details for
Quantity
  • Availability
  • Add to Cart