SY3H3D680D10 Synthonix Corporation

2-(3-Methyl-5-(trifluoromethyl)phenyl)acetic acid

  • MDL number No MDL found
  • Cas Number : 1214352-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54017.html

Safety Information

MSDS https://synthonix.com/54017.html
Availability details for
Quantity
  • Availability
  • Add to Cart