SY3H3D680D1C Synthonix Corporation

Methyl 2-(bromomethyl)-6-cyanobenzoate

  • MDL number No MDL found
  • Cas Number : 1022980-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/53746.html

Safety Information

MSDS https://synthonix.com/53746.html
Availability details for
Quantity
  • Availability
  • Add to Cart