SY3H3D680D1E Synthonix Corporation

1-Methyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • Cas Number : 111635-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54143.html

Safety Information

MSDS https://synthonix.com/54143.html
Availability details for
Quantity
  • Availability
  • Add to Cart