SY3H3D680D22 Synthonix Corporation

3-(Methylamino)-1-phenylpropan-1-one hydrochloride

  • MDL number No MDL found
  • Cas Number : 2538-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54151.html

Safety Information

MSDS https://synthonix.com/54151.html
Availability details for
Quantity
  • Availability
  • Add to Cart