SY3H3D680D23 Synthonix Corporation

4-Methoxy-2,3-dihydro-1H-inden-1-amine hydrochloride

  • Cas Number : 41566-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54152.html

Safety Information

MSDS https://synthonix.com/54152.html
Availability details for
Quantity
  • Availability
  • Add to Cart