SY3H3D680D2D Synthonix Corporation

(S)-Methyl 3-amino-3-phenylpropanoate hydrochloride

  • MDL number No MDL found
  • Cas Number : 144494-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54166.html

Safety Information

MSDS https://synthonix.com/54166.html
Availability details for
Quantity
  • Availability
  • Add to Cart