SY3H3D680D3B Synthonix Corporation

5-Methoxyisoquinoline hydrochloride

  • MDL number No MDL found
  • Cas Number : 1418117-87-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54231.html

Safety Information

MSDS https://synthonix.com/54231.html
Availability details for
Quantity
  • Availability
  • Add to Cart