SY3H3D680D3C Synthonix Corporation

(S)-Methyl 2-amino-2-(3-fluoro-4-(trifluoromethyl)phenyl)acetate hydrochloride

  • MDL number No MDL found
  • Cas Number : 2061996-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54233.html

Safety Information

MSDS https://synthonix.com/54233.html
Availability details for
Quantity
  • Availability
  • Add to Cart