SY3H3D680D41 Synthonix Corporation

4-(1-Methyl-1H-pyrazol-3-yl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 179055-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/54274.html

Safety Information

MSDS https://synthonix.com/54274.html
Availability details for
Quantity
  • Availability
  • Add to Cart