SY3H3D680D42 Synthonix Corporation

2-(2-Methyl-1H-imidazol-1-yl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 914348-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54275.html

Safety Information

MSDS https://synthonix.com/54275.html
Availability details for
Quantity
  • Availability
  • Add to Cart