SY3H3D680D44 Synthonix Corporation

4-(4-Methoxyphenyl)pyrimidin-2-ol

  • MDL number MFCD04050517
  • Cas Number : 674810-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/54279.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/54279.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart