SY3H3D680D45 Synthonix Corporation

Methyl 2-(quinoxalin-6-yl)acetate

  • MDL number No MDL found
  • Cas Number : 1233318-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54280.html

Safety Information

MSDS https://synthonix.com/54280.html
Availability details for
Quantity
  • Availability
  • Add to Cart