SY3H3D680D46 Synthonix Corporation

Methyl 4-(1H-pyrazol-5-yl)benzoate

  • MDL number No MDL found
  • Cas Number : 179057-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54281.html

Safety Information

MSDS https://synthonix.com/54281.html
Availability details for
Quantity
  • Availability
  • Add to Cart