SY3H3D680D47 Synthonix Corporation

2-Methyl-3-(2-nitrovinyl)-1H-indole

  • MDL number No MDL found
  • Cas Number : 2826-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54282.html

Safety Information

MSDS https://synthonix.com/54282.html
Availability details for
Quantity
  • Availability
  • Add to Cart