SY3H3D680D4B Synthonix Corporation

4-(1-Methyl-1H-pyrazol-4-yl)benzoic acid

  • Cas Number : 199678-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54288.html

Safety Information

MSDS https://synthonix.com/54288.html
Availability details for
Quantity
  • Availability
  • Add to Cart