SY3H3D680D4D Synthonix Corporation

2-(2-Methyl-1H-imidazol-1-yl)benzoic acid

  • MDL number No MDL found
  • Cas Number : 159589-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54290.html

Safety Information

MSDS https://synthonix.com/54290.html
Availability details for
Quantity
  • Availability
  • Add to Cart