SY3H3D680D51 Synthonix Corporation

7-Methoxy-2-oxo-1,2,3,4-tetrahydroquinoline-4-carbonitrile

  • Cas Number : 1567360-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54295.html

Safety Information

MSDS https://synthonix.com/54295.html
Availability details for
Quantity
  • Availability
  • Add to Cart