SY3H3D680D52 Synthonix Corporation

Methyl 1-acetyl-1H-indazole-5-carboxylate

  • Cas Number : 239075-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54296.html

Safety Information

MSDS https://synthonix.com/54296.html
Availability details for
Quantity
  • Availability
  • Add to Cart