SY3H3D680E5B Synthonix Corporation

Methyl 3-Bromo-1-methylindole-6-carboxylate

  • MDL number No MDL found
  • Cas Number : 1186663-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54324.html

Safety Information

MSDS https://synthonix.com/54324.html
Availability details for
Quantity
  • Availability
  • Add to Cart