SY3H3D680E5D Synthonix Corporation

Methyl 2-(5-chloro-1H-indol-3-yl)acetate

  • MDL number No MDL found
  • Cas Number : 74339-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/54329.html

Safety Information

MSDS https://synthonix.com/54329.html
Availability details for
Quantity
  • Availability
  • Add to Cart