SY3H3D680E5F Synthonix Corporation

Methyl 5-fluoro-6-methoxy-1H-indole-2-carboxylate

  • MDL number No MDL found
  • Cas Number : 136818-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54334.html

Safety Information

MSDS https://synthonix.com/54334.html
Availability details for
Quantity
  • Availability
  • Add to Cart