SY3H3D680E61 Synthonix Corporation

Methyl 6-chloro-4-methyl-3-oxo-3,4-dihydro-2H-benzo[b][1,4]oxazine-8-carboxylate

  • MDL number No MDL found
  • Cas Number : 141761-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54336.html

Safety Information

MSDS https://synthonix.com/54336.html
Availability details for
Quantity
  • Availability
  • Add to Cart