SY3H3D680E65 Synthonix Corporation

2-(3-Methoxyphenyl)furan

  • MDL number No MDL found
  • Cas Number : 35461-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54341.html

Safety Information

MSDS https://synthonix.com/54341.html
Availability details for
Quantity
  • Availability
  • Add to Cart