SY3H3D680E67 Synthonix Corporation

6-Methoxynaphthalen-1-ol

  • MDL number No MDL found
  • Cas Number : 22604-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54342.html

Safety Information

MSDS https://synthonix.com/54342.html
Availability details for
Quantity
  • Availability
  • Add to Cart