SY3H3D680E6D Synthonix Corporation

Methyl 2-oxo-4-phenylbut-3-enoate

  • Cas Number : 6395-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54350.html

Safety Information

MSDS https://synthonix.com/54350.html
Availability details for
Quantity
  • Availability
  • Add to Cart