SY3H3D680E73 Synthonix Corporation

Methyl 4-(3-hydroxyprop-1-yn-1-yl)benzoate

  • MDL number No MDL found
  • Cas Number : 61266-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54353.html

Safety Information

MSDS https://synthonix.com/54353.html
Availability details for
Quantity
  • Availability
  • Add to Cart