SY3H3D680E7E Synthonix Corporation

4-Methylnaphthalen-1-amine

  • MDL number No MDL found
  • Cas Number : 4523-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54372.html

Safety Information

MSDS https://synthonix.com/54372.html
Availability details for
Quantity
  • Availability
  • Add to Cart