SY3H3D680E88 Synthonix Corporation

6-(4-Methoxyphenoxy)pyridazin-3-amine

  • MDL number No MDL found
  • Cas Number : 121041-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/54394.html

Safety Information

MSDS https://synthonix.com/54394.html
Availability details for
Quantity
  • Availability
  • Add to Cart