SY3H3D680E8E Synthonix Corporation

1-(4-Methoxyphenyl)-1H-pyrrole

  • MDL number No MDL found
  • Cas Number : 5145-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54405.html

Safety Information

MSDS https://synthonix.com/54405.html
Availability details for
Quantity
  • Availability
  • Add to Cart