SY3H3D680E92 Synthonix Corporation

2-(5-Methyl-1H-indol-3-yl)acetic acid

  • Cas Number : 1912-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/54420.html

Safety Information

MSDS https://synthonix.com/54420.html
Availability details for
Quantity
  • Availability
  • Add to Cart