SY3H3D680E9A Synthonix Corporation

4-Methylnaphthalene-1-sulfonamide

  • MDL number No MDL found
  • Cas Number : 10447-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/54426.html

Safety Information

MSDS https://synthonix.com/54426.html
Availability details for
Quantity
  • Availability
  • Add to Cart